Product Details

SNP ID
rs201989257
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:171522648 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCCCTCGTCTGGGTCCTCCACTAC[C/G]GAGAGGGGCTTGGCTGGGATGGGAG
Phenotype
MIM: 605745
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CYBRD1 PubMed Links

Gene Details

Gene
CYBRD1
Gene Name
cytochrome b reductase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001127383.1 293 Missense Mutation CGA,GGA R35G NP_001120855.1
NM_001256909.1 293 Intron NP_001243838.1
NM_024843.3 293 Missense Mutation CGA,GGA R35G NP_079119.3

View Full Product Details