Product Details

SNP ID
rs202003358
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:73700639 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAAGAAATCATAGACGCCCTGCCT[C/G]AGCAGAAGGGAGGTGATAGATGAAA
Phenotype
MIM: 608190
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
NAT8B PubMed Links

Gene Details

Gene
NAT8B
Gene Name
N-acetyltransferase 8B (putative, gene/pseudogene)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016347.2 702 Missense Mutation CAG,GAG Q223E NP_057431.2

View Full Product Details