Product Details

SNP ID
rs200676989
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:50215455 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCTGCTACTCAAGTCCTCAGGGGT[C/T]GTGGGTGAGCCTCACACCAGCCTGG
Phenotype
MIM: 604437
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SLC38A3 PubMed Links

Gene Details

Gene
SLC38A3
Gene Name
solute carrier family 38 member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006841.5 523 Silent Mutation GTC,GTT V123V NP_006832.1
XM_006712954.2 523 Silent Mutation GTC,GTT V123V XP_006713017.1

View Full Product Details