Product Details
- SNP ID
-
hCV191100941
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:71746198 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGACTACAGGATATTCTTCAATTCA[G/A]CATCAATCTGATAATGAAAAAAGAA
- Phenotype
-
MIM: 139200
- Polymorphism
- G/A, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
GC
PubMed Links
Gene Details
- Gene
- GC
- Gene Name
- GC, vitamin D binding protein
There are no transcripts associated with this gene.
View Full Product Details