Product Details

SNP ID
rs199519128
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:51977931 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCCCTGCTGGGCCTGAGTCTGCTG[C/T]TCATGCTCCTACTGTATGTGGGGCT
Phenotype
MIM: 104620
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ABHD14A PubMed Links

Gene Details

Gene
ABHD14A
Gene Name
abhydrolase domain containing 14A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015407.4 240 Missense Mutation CTC,TTC L44F NP_056222.2
Gene
ABHD14A-ACY1
Gene Name
ABHD14A-ACY1 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001316331.1 240 UTR 5 NP_001303260.1
Gene
ABHD14B
Gene Name
abhydrolase domain containing 14B
There are no transcripts associated with this gene.

Gene
ACY1
Gene Name
aminoacylase 1
There are no transcripts associated with this gene.

View Full Product Details