Product Details

SNP ID
rs199523319
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:194687030 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCTGCACATCACTAGCGAGGACCC[A/C]ACTTACACCGTGCTCTACCTGGGCA
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
FAM43A PubMed Links

Gene Details

Gene
FAM43A
Gene Name
family with sequence similarity 43 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153690.4 1138 Silent Mutation CCA,CCC P68P NP_710157.2

View Full Product Details