Product Details

SNP ID
rs199473332
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:38550481 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAAGTGGAGGAGATGGAGGAGCTG[G/A]AGGGTGGGCCAAGGGGTCGGGAGAA
Phenotype
MIM: 600163
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
SCN5A PubMed Links

Gene Details

Gene
SCN5A
Gene Name
sodium voltage-gated channel alpha subunit 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000335.4 6031 Missense Mutation TCC,TTC S1963F NP_000326.2
NM_001099404.1 6031 Missense Mutation TCC,TTC S1964F NP_001092874.1
NM_001099405.1 6031 Missense Mutation TCC,TTC S1946F NP_001092875.1
NM_001160160.1 6031 Missense Mutation TCC,TTC S1931F NP_001153632.1
NM_001160161.1 6031 Missense Mutation TCC,TTC S1910F NP_001153633.1
NM_198056.2 6031 Missense Mutation TCC,TTC S1964F NP_932173.1
XM_011533991.2 6031 Missense Mutation TCC,TTC S1963F XP_011532293.1
XM_017007017.1 6031 Missense Mutation TCC,TTC S1910F XP_016862506.1

View Full Product Details