Product Details

SNP ID
rs201880319
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:98283373 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTAGGTGGCTTCCTCCATGCCTT[A/C]ATTCATGAAGTCCTTATATTCAGAT
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
OR5H2 PubMed Links

Gene Details

Gene
OR5H2
Gene Name
olfactory receptor family 5 subfamily H member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005482.1 486 Missense Mutation TTA,TTC L162F NP_001005482.1

View Full Product Details