Product Details

SNP ID
rs201898990
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:12738140 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATTTGAGCTTGTGTATTTACCACT[G/T]GCTGGGTCAGAGCCTCTCCTTCGGA
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
TMEM40 PubMed Links

Gene Details

Gene
TMEM40
Gene Name
transmembrane protein 40
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001284406.1 635 Silent Mutation GCA,GCC A156A NP_001271335.1
NM_001284407.1 635 Silent Mutation GCA,GCC A110A NP_001271336.1
NM_001284408.1 635 Silent Mutation GCA,GCC A64A NP_001271337.1
NM_018306.3 635 Silent Mutation GCA,GCC A140A NP_060776.2
XM_011533937.2 635 Silent Mutation GCA,GCC A140A XP_011532239.1

View Full Product Details