Product Details

SNP ID
rs201782392
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:46671891 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCTCCTGACTGCCTCACCGGGCGC[A/G]CGACACCACGTAGATGAGAAGTGGC
Phenotype
MIM: 612402
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ALS2CL PubMed Links

Gene Details

Gene
ALS2CL
Gene Name
ALS2 C-terminal like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001190707.1 2773 Missense Mutation CGC,TGC R893C NP_001177636.1
NM_147129.4 2773 Missense Mutation CGC,TGC R893C NP_667340.2
XM_005265025.1 2773 Missense Mutation CGC,TGC R408C XP_005265082.1
XM_006713091.2 2773 Intron XP_006713154.1
XM_006713093.3 2773 Intron XP_006713156.1
XM_006713094.3 2773 Intron XP_006713157.1
XM_011533572.2 2773 Intron XP_011531874.1
XM_017006120.1 2773 Intron XP_016861609.1

View Full Product Details