Product Details

SNP ID
rs201853202
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:187726780 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGAGGTTGCCCTTGTAGCGGAAGGA[C/G]GCCTGGCAGCGGTCACACTTGTAGG
Phenotype
MIM: 109565
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
BCL6 PubMed Links

Gene Details

Gene
BCL6
Gene Name
B-cell CLL/lymphoma 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130845.1 2024 Silent Mutation GCC,GCG A553A NP_001124317.1
NM_001134738.1 2024 Intron NP_001128210.1
NM_001706.4 2024 Silent Mutation GCC,GCG A553A NP_001697.2
XM_005247694.3 2024 Silent Mutation GCC,GCG A553A XP_005247751.1
XM_011513062.2 2024 Intron XP_011511364.1
Gene
LOC100131635
Gene Name
hCG1645011-like
There are no transcripts associated with this gene.

View Full Product Details