Product Details

SNP ID
rs201822269
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:27717717 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGGGCTCCGGGAAGAAGGATTGAA[C/T]GCCGTACCGACCTCCAGGGACAATC
Phenotype
MIM: 604615
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
EOMES PubMed Links

Gene Details

Gene
EOMES
Gene Name
eomesodermin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278182.1 1835 Missense Mutation ATT,GTT I491V NP_001265111.1
NM_001278183.1 1835 Missense Mutation ATT,GTT I196V NP_001265112.1
NM_005442.3 1835 Missense Mutation ATT,GTT I472V NP_005433.2
XM_005265510.3 1835 Intron XP_005265567.1

View Full Product Details