Product Details
- SNP ID
-
rs201924367
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:138010333 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACATGTGGAGCACCCAGGACCTGTA[C/T]GACAACCCCGTCACCTCCGTGTTCC
- Phenotype
-
MIM: 609210
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CLDN18
PubMed Links
Gene Details
- Gene
- CLDN18
- Gene Name
- claudin 18
View Full Product Details