Product Details

SNP ID
rs201894452
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:151213533 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTGACTGGCAGCTGTAATGAGCTT[C/T]GGTCTGACTCTTTGTGTAGGGGATT
Phenotype
MIM: 611318 MIM: 610116
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GPR171 PubMed Links

Gene Details

Gene
GPR171
Gene Name
G protein-coupled receptor 171
There are no transcripts associated with this gene.

Gene
MED12L
Gene Name
mediator complex subunit 12 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_053002.5 1072 Intron NP_443728.3
XM_006713487.3 1072 Intron XP_006713550.1
XM_011512390.2 1072 Intron XP_011510692.1
XM_011512394.2 1072 Intron XP_011510696.1
XM_011512399.2 1072 Intron XP_011510701.1
XM_017005676.1 1072 Intron XP_016861165.1
XM_017005677.1 1072 Intron XP_016861166.1
XM_017005678.1 1072 Intron XP_016861167.1
XM_017005679.1 1072 Intron XP_016861168.1
XM_017005680.1 1072 Intron XP_016861169.1
XM_017005681.1 1072 Intron XP_016861170.1
Gene
P2RY14
Gene Name
purinergic receptor P2Y14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001081455.1 1072 Missense Mutation AAA,GAA K262E NP_001074924.1
NM_014879.3 1072 Missense Mutation AAA,GAA K262E NP_055694.3
XM_005247922.3 1072 Missense Mutation AAA,GAA K262E XP_005247979.1
XM_005247923.3 1072 Missense Mutation AAA,GAA K262E XP_005247980.1
XM_011513340.2 1072 Missense Mutation AAA,GAA K262E XP_011511642.1
XM_017007583.1 1072 Missense Mutation AAA,GAA K262E XP_016863072.1

View Full Product Details