Product Details

SNP ID
rs202133841
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:5187852 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGGGCTGGTGCTCCTCCGGCTTG[C/G]CCTCCATGGAGTATTGTGGCTCGTC
Phenotype
MIM: 616596 MIM: 607673
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ARL8B PubMed Links

Gene Details

Gene
ARL8B
Gene Name
ADP ribosylation factor like GTPase 8B
There are no transcripts associated with this gene.

Gene
EDEM1
Gene Name
ER degradation enhancing alpha-mannosidase like protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014674.2 109 Missense Mutation GCC,GGC A16G NP_055489.1
XM_011534271.2 109 Intron XP_011532573.1
XM_011534272.2 109 UTR 5 XP_011532574.1

View Full Product Details