Product Details

SNP ID
rs201673223
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:151328349 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTTTTGGACTTTCTATAAGAATCA[C/T]ATACTTTTTTTGCAATAACCACATA
Phenotype
MIM: 611318 MIM: 600515 MIM: 606380
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MED12L PubMed Links

Gene Details

Gene
MED12L
Gene Name
mediator complex subunit 12 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_053002.5 728 Intron NP_443728.3
XM_006713487.3 728 Intron XP_006713550.1
XM_011512390.2 728 Intron XP_011510692.1
XM_011512394.2 728 Intron XP_011510696.1
XM_011512399.2 728 Intron XP_011510701.1
XM_017005676.1 728 Intron XP_016861165.1
XM_017005677.1 728 Intron XP_016861166.1
XM_017005678.1 728 Intron XP_016861167.1
XM_017005679.1 728 Intron XP_016861168.1
XM_017005680.1 728 Intron XP_016861169.1
XM_017005681.1 728 Intron XP_016861170.1
Gene
P2RY12
Gene Name
purinergic receptor P2Y12
There are no transcripts associated with this gene.

Gene
P2RY13
Gene Name
purinergic receptor P2Y13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_176894.2 728 Missense Mutation TAT,TGT Y236C NP_795713.2
XM_006713664.1 728 Intron XP_006713727.1

View Full Product Details