Product Details

SNP ID
rs201139758
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:48433868 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTCGAGCCACCATGAACACTTTCT[C/G]AGCCTGCAAAGAGAAAACCGGAGGC
Phenotype
MIM: 601053 MIM: 615808
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CCDC51 PubMed Links

Gene Details

Gene
CCDC51
Gene Name
coiled-coil domain containing 51
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256964.1 783 Missense Mutation CAG,GAG Q106E NP_001243893.1
NM_001256965.2 783 UTR 5 NP_001243894.1
NM_001256966.2 783 UTR 5 NP_001243895.1
NM_001256967.2 783 UTR 5 NP_001243896.1
NM_001256968.2 783 UTR 5 NP_001243897.1
NM_001256969.2 783 UTR 5 NP_001243898.1
NM_024661.4 783 Missense Mutation CAG,GAG Q106E NP_078937.3
XM_011534113.2 783 Missense Mutation CAG,GAG Q126E XP_011532415.1
Gene
PLXNB1
Gene Name
plexin B1
There are no transcripts associated with this gene.

Gene
TMA7
Gene Name
translation machinery associated 7 homolog
There are no transcripts associated with this gene.

View Full Product Details