Product Details
- SNP ID
-
rs201708744
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:51958349 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAGAGGTAGACTTGGCCGTCTCTAG[A/G]CTGGAGAGAGGGATATAGAGGGGAG
- Phenotype
-
MIM: 606917
MIM: 607726
MIM: 608503
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
GPR62
PubMed Links
Gene Details
- Gene
- GPR62
- Gene Name
- G protein-coupled receptor 62
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_080865.3 |
1277 |
Intron |
|
|
NP_543141.3 |
- Gene
- PARP3
- Gene Name
- poly(ADP-ribose) polymerase family member 3
There are no transcripts associated with this gene.
- Gene
- PCBP4
- Gene Name
- poly(rC) binding protein 4
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001174100.1 |
1277 |
Silent Mutation |
TGC,TGT |
C308C |
NP_001167571.1 |
NM_020418.3 |
1277 |
Silent Mutation |
TGC,TGT |
C265C |
NP_065151.2 |
NM_033008.2 |
1277 |
Silent Mutation |
TGC,TGT |
C308C |
NP_127501.1 |
NM_033010.2 |
1277 |
Silent Mutation |
TGC,TGT |
C308C |
NP_127503.1 |
XM_005265330.2 |
1277 |
Silent Mutation |
TGC,TGT |
C331C |
XP_005265387.1 |
XM_005265331.2 |
1277 |
Silent Mutation |
TGC,TGT |
C308C |
XP_005265388.1 |
XM_005265332.2 |
1277 |
Silent Mutation |
TGC,TGT |
C288C |
XP_005265389.1 |
XM_006713268.2 |
1277 |
Silent Mutation |
TGC,TGT |
C308C |
XP_006713331.1 |
XM_006713269.1 |
1277 |
Silent Mutation |
TGC,TGT |
C308C |
XP_006713332.1 |
XM_006713270.1 |
1277 |
Silent Mutation |
TGC,TGT |
C265C |
XP_006713333.1 |
XM_006713271.2 |
1277 |
Intron |
|
|
XP_006713334.1 |
XM_017006916.1 |
1277 |
Silent Mutation |
TGC,TGT |
C331C |
XP_016862405.1 |
XM_017006917.1 |
1277 |
Intron |
|
|
XP_016862406.1 |
XM_017006918.1 |
1277 |
Intron |
|
|
XP_016862407.1 |
XM_017006919.1 |
1277 |
Intron |
|
|
XP_016862408.1 |
XM_017006920.1 |
1277 |
Intron |
|
|
XP_016862409.1 |
View Full Product Details