Product Details

SNP ID
rs200815644
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:196577682 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAAAACGGGCCCCCATGCAGTGCC[A/G]AGGTTATGTGGCATTGCTGGGCAGT
Phenotype
MIM: 609112 MIM: 615110
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FBXO45 PubMed Links

Gene Details

Gene
FBXO45
Gene Name
F-box protein 45
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001105573.1 679 Missense Mutation CAA,CGA Q183R NP_001099043.1
Gene
WDR53
Gene Name
WD repeat domain 53
There are no transcripts associated with this gene.

View Full Product Details