Product Details
- SNP ID
-
rs201106888
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:184711041 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAGTGCTGCGGTTCCTTCTTATGCA[A/G]TTTGGCCACGAACCCCAGGACTTCC
- Phenotype
-
MIM: 609267
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
MAGEF1
PubMed Links
Gene Details
- Gene
- MAGEF1
- Gene Name
- MAGE family member F1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_022149.4 |
1008 |
Silent Mutation |
CTG,TTG |
L261L |
NP_071432.2 |
View Full Product Details