Product Details

SNP ID
rs201487546
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:50617640 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGCACCCGGCGGACCCGGCTTGGG[C/T]GGTGCTCCGGGGGGGCGGCGGGAGC
Phenotype
MIM: 602441 MIM: 602130
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CISH PubMed Links

Gene Details

Gene
CISH
Gene Name
cytokine inducible SH2 containing protein
There are no transcripts associated with this gene.

Gene
MAPKAPK3
Gene Name
mitogen-activated protein kinase-activated protein kinase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001243925.1 238 Silent Mutation GGC,GGT G25G NP_001230854.1
NM_001243926.1 238 Silent Mutation GGC,GGT G25G NP_001230855.1
NM_004635.4 238 Silent Mutation GGC,GGT G25G NP_004626.1

View Full Product Details