Product Details

SNP ID
rs201763792
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:154338201 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATAGGTAGAAAGAGATAGGGTTTTC[C/T]CTGTAGGTGCATGGAGAGACACAGT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GPR149 PubMed Links

Gene Details

Gene
GPR149
Gene Name
G protein-coupled receptor 149
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001038705.1 1794 Missense Mutation GAG,GGG E565G NP_001033794.1

View Full Product Details