Product Details

SNP ID
rs201609649
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:28324128 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTAAAGGCAAAGTTTTAGTTTTAG[A/T]TTCCCCAAATGCTGTCTCACTTGAT
Phenotype
MIM: 609916 MIM: 615166
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
AZI2 PubMed Links

Gene Details

Gene
AZI2
Gene Name
5-azacytidine induced 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001134432.1 1165 Intron NP_001127904.1
NM_001134433.1 1165 Intron NP_001127905.1
NM_001271650.1 1165 Missense Mutation ACT,TCT T253S NP_001258579.1
NM_022461.4 1165 Missense Mutation ACT,TCT T365S NP_071906.1
XM_005265392.2 1165 Missense Mutation ACT,TCT T365S XP_005265449.1
XM_011534009.2 1165 Missense Mutation ACT,TCT T368S XP_011532311.1
XM_011534010.2 1165 Intron XP_011532312.1
XM_011534011.2 1165 Intron XP_011532313.1
Gene
CMC1
Gene Name
C-X9-C motif containing 1
There are no transcripts associated with this gene.

View Full Product Details