Product Details
- SNP ID
-
rs201507517
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:31664209 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTCAATGTCCCCCAGCCGCAGGTAT[C/T]GGGTCACCTCCCGCCAGAGGTTCCT
- Phenotype
-
MIM: 606738
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
OSBPL10
PubMed Links
Gene Details
- Gene
- OSBPL10
- Gene Name
- oxysterol binding protein like 10
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001174060.1 |
1963 |
Missense Mutation |
CAA,CGA |
Q643R |
NP_001167531.1 |
NM_017784.4 |
1963 |
Missense Mutation |
CAA,CGA |
Q707R |
NP_060254.2 |
XM_005264843.3 |
1963 |
Missense Mutation |
CAA,CGA |
Q707R |
XP_005264900.1 |
XM_005264844.4 |
1963 |
Missense Mutation |
CAA,CGA |
Q643R |
XP_005264901.1 |
XM_005264845.1 |
1963 |
Missense Mutation |
CAA,CGA |
Q371R |
XP_005264902.1 |
XM_011533326.2 |
1963 |
Missense Mutation |
CAA,CGA |
Q570R |
XP_011531628.1 |
XM_017005667.1 |
1963 |
Missense Mutation |
CAA,CGA |
Q570R |
XP_016861156.1 |
XM_017005668.1 |
1963 |
Missense Mutation |
CAA,CGA |
Q570R |
XP_016861157.1 |
XM_017005669.1 |
1963 |
Missense Mutation |
CAA,CGA |
Q534R |
XP_016861158.1 |
XM_017005670.1 |
1963 |
Missense Mutation |
CAA,CGA |
Q489R |
XP_016861159.1 |
View Full Product Details