Product Details

SNP ID
rs201754591
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:150742351 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTAGGCAAAGTTCTCGGCTTGCTT[G/T]CGGGTGCCAATGAGCAGGACGATGG
Phenotype
MIM: 602213
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
LOC101928105 PubMed Links

Gene Details

Gene
LOC101928105
Gene Name
uncharacterized LOC101928105
There are no transcripts associated with this gene.

Gene
SIAH2
Gene Name
siah E3 ubiquitin protein ligase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005067.5 1392 Silent Mutation CGA,CGC R255R NP_005058.3

View Full Product Details