Product Details

SNP ID
rs201721015
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:187370919 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCTTTGGCCAAAGGTGCCAGAAGT[A/G]CTCCTGGTCCCAATATGAGATGCCT
Phenotype
MIM: 609350
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RTP4 PubMed Links

Gene Details

Gene
RTP4
Gene Name
receptor transporter protein 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022147.2 349 Missense Mutation TAC,TGC Y96C NP_071430.2

View Full Product Details