Product Details

SNP ID
rs201112544
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:50271402 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGGGTGGCAGCAGACCTCATGGGA[C/T]GAGACTTTACCATCTTTCGCAGCCT
Phenotype
MIM: 601281
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LSMEM2 PubMed Links

Gene Details

Gene
LSMEM2
Gene Name
leucine rich single-pass membrane protein 2
There are no transcripts associated with this gene.

Gene
MIR6872
Gene Name
microRNA 6872
There are no transcripts associated with this gene.

Gene
SEMA3B
Gene Name
semaphorin 3B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005914.2 878 Nonsense Mutation CGA,TGA R196* NP_001005914.1
NM_001290060.1 878 Nonsense Mutation CGA,TGA R196* NP_001276989.1
NM_001290061.1 878 Nonsense Mutation CGA,TGA R196* NP_001276990.1
NM_001290062.1 878 Intron NP_001276991.1
NM_001290063.1 878 Intron NP_001276992.1
NM_004636.3 878 Nonsense Mutation CGA,TGA R196* NP_004627.1
Gene
SEMA3B-AS1
Gene Name
SEMA3B antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

View Full Product Details