Product Details

SNP ID
rs201198335
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:13317772 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCATTGGGAGATGTGGGTGATGAG[A/G]CAGCGAAATCTAGGGTGGGAAGAGA
Phenotype
MIM: 607703
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NUP210 PubMed Links

Gene Details

Gene
NUP210
Gene Name
nucleoporin 210
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024923.3 5538 Missense Mutation GCC,GTC A1858V NP_079199.2
XM_005264994.3 5538 Intron XP_005265051.1
XM_011533538.2 5538 Intron XP_011531840.1
XM_017006018.1 5538 Missense Mutation GCC,GTC A986V XP_016861507.1
XM_017006019.1 5538 Missense Mutation GCC,GTC A975V XP_016861508.1
XM_017006020.1 5538 Missense Mutation GCC,GTC A963V XP_016861509.1
XM_017006021.1 5538 Intron XP_016861510.1

View Full Product Details