Product Details
- SNP ID
-
rs201599722
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:123284699 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CACGCCCCGGCACTCCAGCTGGTAC[A/G]TGTTGGCAGCCAGCACCTGGTACAT
- Phenotype
-
MIM: 600293
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ADCY5
PubMed Links
Gene Details
- Gene
- ADCY5
- Gene Name
- adenylate cyclase 5
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001199642.1 |
2791 |
Missense Mutation |
ACG,ATG |
T882M |
NP_001186571.1 |
NM_183357.2 |
2791 |
Missense Mutation |
ACG,ATG |
T1232M |
NP_899200.1 |
XM_005247077.3 |
2791 |
Missense Mutation |
ACG,ATG |
T1257M |
XP_005247134.1 |
XM_005247078.1 |
2791 |
Missense Mutation |
ACG,ATG |
T907M |
XP_005247135.1 |
XM_006713483.1 |
2791 |
Missense Mutation |
ACG,ATG |
T890M |
XP_006713546.1 |
XM_006713484.1 |
2791 |
Missense Mutation |
ACG,ATG |
T816M |
XP_006713547.1 |
XM_011512359.2 |
2791 |
Missense Mutation |
ACG,ATG |
T924M |
XP_011510661.1 |
XM_011512360.2 |
2791 |
Missense Mutation |
ACG,ATG |
T894M |
XP_011510662.1 |
XM_011512361.1 |
2791 |
Missense Mutation |
ACG,ATG |
T816M |
XP_011510663.1 |
XM_017005638.1 |
2791 |
Missense Mutation |
ACG,ATG |
T891M |
XP_016861127.1 |
XM_017005639.1 |
2791 |
Missense Mutation |
ACG,ATG |
T891M |
XP_016861128.1 |
XM_017005640.1 |
2791 |
Missense Mutation |
ACG,ATG |
T816M |
XP_016861129.1 |
View Full Product Details