Product Details

SNP ID
rs200825196
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:50300677 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGAGGGCTTTGGGGTGGCTGGTGC[A/G]GCGGACACAGCGGCCATGGCCGGAG
Phenotype
MIM: 607071 MIM: 604038 MIM: 602725 MIM: 607073
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HYAL1 PubMed Links

Gene Details

Gene
HYAL1
Gene Name
hyaluronoglucosaminidase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033159.3 1952 Missense Mutation CGC,TGC R372C NP_149349.2
NM_153281.1 1952 Missense Mutation CGC,TGC R372C NP_695013.1
NM_153282.2 1952 Missense Mutation CGC,TGC R342C NP_695014.1
NM_153283.2 1952 Missense Mutation CGC,TGC R190C NP_695015.1
NM_153285.2 1952 Missense Mutation CGC,TGC R113C NP_695017.1
XM_011533667.2 1952 Missense Mutation CGC,TGC R372C XP_011531969.1
XM_011533668.2 1952 Missense Mutation CGC,TGC R372C XP_011531970.1
XM_011533669.2 1952 Missense Mutation CGC,TGC R372C XP_011531971.1
Gene
HYAL3
Gene Name
hyaluronoglucosaminidase 3
There are no transcripts associated with this gene.

Gene
IFRD2
Gene Name
interferon related developmental regulator 2
There are no transcripts associated with this gene.

Gene
NAT6
Gene Name
N-acetyltransferase 6
There are no transcripts associated with this gene.

View Full Product Details