Product Details
- SNP ID
-
rs201046832
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:78600143 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATTCTTCATTATTATCTTCTCCTCT[A/T]TCATATCCTCCAAGTACCTGCATTT
- Phenotype
-
MIM: 602430
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ROBO1
PubMed Links
Gene Details
- Gene
- ROBO1
- Gene Name
- roundabout guidance receptor 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001145845.1 |
5070 |
Missense Mutation |
GAA,GAT |
E1537D |
NP_001139317.1 |
NM_002941.3 |
5070 |
Missense Mutation |
GAA,GAT |
E1637D |
NP_002932.1 |
NM_133631.3 |
5070 |
Missense Mutation |
GAA,GAT |
E1592D |
NP_598334.2 |
XM_006713277.2 |
5070 |
Missense Mutation |
GAA,GAT |
E1601D |
XP_006713340.1 |
XM_011533976.1 |
5070 |
Missense Mutation |
GAA,GAT |
E1640D |
XP_011532278.1 |
XM_011533977.2 |
5070 |
Missense Mutation |
GAA,GAT |
E1640D |
XP_011532279.1 |
XM_011533978.1 |
5070 |
Missense Mutation |
GAA,GAT |
E1631D |
XP_011532280.1 |
XM_011533979.1 |
5070 |
Missense Mutation |
GAA,GAT |
E1585D |
XP_011532281.1 |
XM_011533980.1 |
5070 |
Missense Mutation |
GAA,GAT |
E1576D |
XP_011532282.1 |
XM_017006982.1 |
5070 |
Missense Mutation |
GAA,GAT |
E1612D |
XP_016862471.1 |
XM_017006983.1 |
5070 |
Missense Mutation |
GAA,GAT |
E1598D |
XP_016862472.1 |
XM_017006984.1 |
5070 |
Missense Mutation |
GAA,GAT |
E1582D |
XP_016862473.1 |
XM_017006985.1 |
5070 |
Missense Mutation |
GAA,GAT |
E1521D |
XP_016862474.1 |
View Full Product Details