Product Details

SNP ID
rs201667583
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:143120881 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCCCTCGGCTGCTCCAGGCTGCGC[C/T]TGCAGCCGCGCCGCGTGCCCTGCTC
Phenotype
MIM: 603798
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CHST2 PubMed Links

Gene Details

Gene
CHST2
Gene Name
carbohydrate sulfotransferase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004267.4 1004 Missense Mutation CCT,CTT P22L NP_004258.2

View Full Product Details