Product Details

SNP ID
rs201359879
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:196708565 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAAGTTTCGAACTGGCATAATGCG[C/G]TGGCGAATTTTCCCCTTGATTTCAC
Phenotype
MIM: 615586 MIM: 610276
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CEP19 PubMed Links

Gene Details

Gene
CEP19
Gene Name
centrosomal protein 19
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032898.4 265 Missense Mutation CAC,CAG H35Q NP_116287.2
XM_005269370.4 265 Missense Mutation CAC,CAG H35Q XP_005269427.1
XM_011513246.2 265 Missense Mutation CAC,CAG H35Q XP_011511548.1
Gene
PIGX
Gene Name
phosphatidylinositol glycan anchor biosynthesis class X
There are no transcripts associated with this gene.

View Full Product Details