Product Details

SNP ID
rs201135546
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:156544317 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCTTGAAAAGGATACACTGTGGGG[C/T]TGAAGTTCTTCAATATGAAGAAGGA
Phenotype
MIM: 601141 MIM: 606213
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KCNAB1 PubMed Links

Gene Details

Gene
KCNAB1
Gene Name
potassium voltage-gated channel subfamily A member regulatory beta subunit 1
There are no transcripts associated with this gene.

Gene
SSR3
Gene Name
signal sequence receptor subunit 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308197.1 632 Missense Mutation AAC,AGC N174S NP_001295126.1
NM_001308204.1 632 Missense Mutation AAC,AGC N109S NP_001295133.1
NM_001308205.1 632 Missense Mutation AAC,AGC N109S NP_001295134.1
NM_007107.4 632 Missense Mutation AAC,AGC N161S NP_009038.1

View Full Product Details