Product Details

SNP ID
rs201015334
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:122565915 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAAATACTTGTTGACGAAAAACCT[A/G]TGCCCATTTTCCTGGTACCCACTGA
Phenotype
MIM: 613143 MIM: 612066 MIM: 612065
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DTX3L PubMed Links

Gene Details

Gene
DTX3L
Gene Name
deltex E3 ubiquitin ligase 3L
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138287.3 333 Missense Mutation ATG,GTG M82V NP_612144.1
Gene
PARP15
Gene Name
poly(ADP-ribose) polymerase family member 15
There are no transcripts associated with this gene.

Gene
PARP9
Gene Name
poly(ADP-ribose) polymerase family member 9
There are no transcripts associated with this gene.

View Full Product Details