Product Details

SNP ID
rs201224636
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:40532101 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCCTCTGAAGAAACAGAGTTGCAC[A/G]GAATGCCAGTAGGAGGACTTCTCAG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF621 PubMed Links

Gene Details

Gene
ZNF621
Gene Name
zinc finger protein 621
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098414.2 617 Missense Mutation AGA,GGA R111G NP_001091884.1
NM_001287245.1 617 Missense Mutation AGA,GGA R111G NP_001274174.1
NM_198484.4 617 Missense Mutation AGA,GGA R111G NP_940886.1
XM_005265079.3 617 Missense Mutation AGA,GGA R103G XP_005265136.1
XM_005265080.3 617 Missense Mutation AGA,GGA R76G XP_005265137.1
XM_006713114.3 617 Missense Mutation AGA,GGA R103G XP_006713177.1
XM_011533610.2 617 Missense Mutation AGA,GGA R103G XP_011531912.1
XM_017006226.1 617 Missense Mutation AGA,GGA R76G XP_016861715.1

View Full Product Details