Product Details

SNP ID
rs200947739
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:49001552 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTGAAGAGTATGAAGAGGCAATGA[C/G]CACTATGCAGGTCAGCCAGCTGGAC
Phenotype
MIM: 614584 MIM: 606031
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
P4HTM PubMed Links

Gene Details

Gene
P4HTM
Gene Name
prolyl 4-hydroxylase, transmembrane
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_177938.2 900 Missense Mutation ACC,AGC T184S NP_808807.2
NM_177939.2 900 Missense Mutation ACC,AGC T184S NP_808808.1
Gene
WDR6
Gene Name
WD repeat domain 6
There are no transcripts associated with this gene.

View Full Product Details