Product Details

SNP ID
rs201137978
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:49859574 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTCCTGTCAGTGGCTGGGGTGACA[C/T]TCCCATCAGTGGCTGGAGTGATGCT
Phenotype
MIM: 614993 MIM: 605958
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CAMKV PubMed Links

Gene Details

Gene
CAMKV
Gene Name
CaM kinase like vesicle associated
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320147.1 1636 Missense Mutation AAT,AGT N386S NP_001307076.1
NM_024046.4 1636 Missense Mutation AAT,AGT N417S NP_076951.2
Gene
TRAIP
Gene Name
TRAF interacting protein
There are no transcripts associated with this gene.

View Full Product Details