Product Details

SNP ID
rs201363121
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:128069494 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCCCAGGTACATCCCCACAGCCGC[A/G]GCCTTTGGTGGGCTGTGCATCGGGG
Phenotype
MIM: 603449 MIM: 609213
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RUVBL1 PubMed Links

Gene Details

Gene
RUVBL1
Gene Name
RuvB like AAA ATPase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001319084.1 1447 Intron NP_001306013.1
NM_001319086.1 1447 Intron NP_001306015.1
NM_003707.2 1447 Intron NP_003698.1
XM_005247841.3 1447 Intron XP_005247898.1
XM_011513248.1 1447 Intron XP_011511550.1
XM_011513249.2 1447 Intron XP_011511551.1
XM_017007356.1 1447 Intron XP_016862845.1
XM_017007357.1 1447 Intron XP_016862846.1
Gene
RUVBL1-AS1
Gene Name
RUVBL1 antisense RNA 1
There are no transcripts associated with this gene.

Gene
SEC61A1
Gene Name
Sec61 translocon alpha 1 subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013336.3 1447 Silent Mutation GCA,GCG A421A NP_037468.1

View Full Product Details