Product Details

SNP ID
rs201415238
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:180606262 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAATGTCTTTTTTAGTGTGAAGAT[C/T]GGAGTTGACTATTTTAAAGTTGGAC
Phenotype
MIM: 613798
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCDC39 PubMed Links

Gene Details

Gene
CCDC39
Gene Name
coiled-coil domain containing 39
There are no transcripts associated with this gene.

Gene
TTC14
Gene Name
tetratricopeptide repeat domain 14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042601.2 1071 Silent Mutation ATC,ATT I313I NP_001036066.1
NM_001288582.1 1071 Silent Mutation ATC,ATT I313I NP_001275511.1
NM_133462.3 1071 Silent Mutation ATC,ATT I313I NP_597719.1
XM_011512439.2 1071 Silent Mutation ATC,ATT I63I XP_011510741.1
XM_017005739.1 1071 Silent Mutation ATC,ATT I261I XP_016861228.1
XM_017005740.1 1071 Silent Mutation ATC,ATT I313I XP_016861229.1

View Full Product Details