Product Details

SNP ID
rs200093616
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:38138862 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTGGACCGCGCTGGCGGAGGAGAT[A/G]GACTTTGAGTACTTGGAGATCCGGC
Phenotype
MIM: 604054 MIM: 602170
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ACAA1 PubMed Links

Gene Details

Gene
ACAA1
Gene Name
acetyl-CoA acyltransferase 1
There are no transcripts associated with this gene.

Gene
MYD88
Gene Name
myeloid differentiation primary response 88
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001172566.1 385 Missense Mutation ATA,ATG I67M NP_001166037.1
NM_001172567.1 385 Missense Mutation ATA,ATG I67M NP_001166038.1
NM_001172568.1 385 Missense Mutation ATA,ATG I67M NP_001166039.1
NM_001172569.1 385 Missense Mutation ATA,ATG I67M NP_001166040.1
NM_002468.4 385 Missense Mutation ATA,ATG I67M NP_002459.2
XM_005265172.1 385 Missense Mutation ATA,ATG I67M XP_005265229.1
XM_006713170.1 385 Missense Mutation ATA,ATG I67M XP_006713233.1

View Full Product Details