Product Details

SNP ID
rs200351516
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:42258116 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGTCCTCTAGGAGGGGTACTCATA[C/T]TCCTCGGCACTGCGACGGCCAAAAT
Phenotype
MIM: 118440
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCK PubMed Links

Gene Details

Gene
CCK
Gene Name
cholecystokinin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000729.5 1224 Silent Mutation GAA,GAG E110E NP_000720.1
NM_001174138.2 1224 Silent Mutation GAA,GAG E110E NP_001167609.1

View Full Product Details