Product Details

SNP ID
rs200611775
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:40530316 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGAGATTCTGAGAGGCATCAGTCAA[A/G]GTGAGTATGAGAATCCAGTGGTGAA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF621 PubMed Links

Gene Details

Gene
ZNF621
Gene Name
zinc finger protein 621
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098414.2 545 Missense Mutation AGT,GGT S87G NP_001091884.1
NM_001287245.1 545 Missense Mutation AGT,GGT S87G NP_001274174.1
NM_198484.4 545 Missense Mutation AGT,GGT S87G NP_940886.1
XM_005265079.3 545 Missense Mutation AGT,GGT S79G XP_005265136.1
XM_005265080.3 545 Missense Mutation AGT,GGT S52G XP_005265137.1
XM_006713114.3 545 Missense Mutation AGT,GGT S79G XP_006713177.1
XM_011533610.2 545 Missense Mutation AGT,GGT S79G XP_011531912.1
XM_017006226.1 545 Missense Mutation AGT,GGT S52G XP_016861715.1

View Full Product Details