Product Details

SNP ID
rs200078271
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:133934740 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGATGAGGCCTGCCGCCTTCTGCAC[A/G]TTGTACTCCTTGTTCTTCTTCACCC
Phenotype
MIM: 601460
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C3orf36 PubMed Links

Gene Details

Gene
C3orf36
Gene Name
chromosome 3 open reading frame 36
There are no transcripts associated with this gene.

Gene
SLCO2A1
Gene Name
solute carrier organic anion transporter family member 2A1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005630.2 2179 Silent Mutation AAC,AAT N635N NP_005621.2
XM_017007075.1 2179 Silent Mutation AAC,AAT N645N XP_016862564.1
XM_017007076.1 2179 Intron XP_016862565.1
XM_017007077.1 2179 Silent Mutation AAC,AAT N467N XP_016862566.1

View Full Product Details