Product Details

SNP ID
rs200438173
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:23807401 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAGCATGAGCAAAAACTCCAAACT[C/T]CTCTCCACCAGCGCCAAGAGGTACT
Phenotype
MIM: 602916
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
UBE2E1 PubMed Links

Gene Details

Gene
UBE2E1
Gene Name
ubiquitin conjugating enzyme E2 E1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001202476.1 361 Intron NP_001189405.1
NM_003341.4 361 Silent Mutation CTC,CTT L44L NP_003332.1
NM_182666.2 361 Silent Mutation CTC,CTT L44L NP_872607.1
XM_005265431.4 361 Intron XP_005265488.1
Gene
UBE2E1-AS1
Gene Name
UBE2E1 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details