Product Details

SNP ID
rs199617465
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:31989389 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCACATTGGAGATTTTTGCTTCCAG[A/G]AAATTGGGAAAGATATTCATGACTT
Phenotype
MIM: 606738
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OSBPL10 PubMed Links

Gene Details

Gene
OSBPL10
Gene Name
oxysterol binding protein like 10
There are no transcripts associated with this gene.

Gene
ZNF860
Gene Name
zinc finger protein 860
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001137674.2 859 Missense Mutation AAA,GAA K104E NP_001131146.2
XM_017006299.1 859 Intron XP_016861788.1

View Full Product Details