Product Details

SNP ID
rs199959056
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:39265467 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAGAGAAGGAGCAATGCATCTCCA[C/T]CACTCGTGTGGTAAGTAAAATTGCT
Phenotype
MIM: 601470
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CX3CR1 PubMed Links

Gene Details

Gene
CX3CR1
Gene Name
C-X3-C motif chemokine receptor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001171171.1 1135 Missense Mutation GAT,GGT D348G NP_001164642.1
NM_001171172.1 1135 Missense Mutation GAT,GGT D348G NP_001164643.1
NM_001171174.1 1135 Missense Mutation GAT,GGT D380G NP_001164645.1
NM_001337.3 1135 Missense Mutation GAT,GGT D348G NP_001328.1

View Full Product Details