Product Details

SNP ID
rs200558767
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:3070239 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAAGTGACAGTCAAAACACAGAAT[C/T]CTCCAGGGTCTCAACTCCAGGCTTC
Phenotype
MIM: 147851
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CNTN4-AS1 PubMed Links

Gene Details

Gene
CNTN4-AS1
Gene Name
CNTN4 antisense RNA 1
There are no transcripts associated with this gene.

Gene
IL5RA
Gene Name
interleukin 5 receptor subunit alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000564.4 1824 Missense Mutation AAT,GAT N417D NP_000555.2
NM_001243099.1 1824 UTR 3 NP_001230028.1
NM_175724.2 1824 Intron NP_783851.1
NM_175725.2 1824 Intron NP_783852.1
NM_175726.3 1824 Missense Mutation AAT,GAT N417D NP_783853.1
NM_175727.2 1824 Intron NP_783854.1
NM_175728.2 1824 Intron NP_783855.1
XM_011533677.2 1824 Missense Mutation AAT,GAT N436D XP_011531979.1
XM_011533678.2 1824 Missense Mutation AAT,GAT N436D XP_011531980.1

View Full Product Details