Product Details
- SNP ID
-
rs199842485
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:133805956 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGGAGCTGCAGCAGACGGACCCAAC[C/G]CTGTTGTCAGTAGTGGTGGCGGTTC
- Phenotype
-
MIM: 616883
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
SRPRB
PubMed Links
Gene Details
- Gene
- SRPRB
- Gene Name
- SRP receptor beta subunit
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_021203.3 |
393 |
Silent Mutation |
ACC,ACG |
T36T |
NP_067026.3 |
View Full Product Details