Product Details

SNP ID
rs200373793
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:50301045 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCACCACTCCAGCTGCCCCCTGGGC[C/G]GCACTCTCCCCCAGGCTGTGCTCCA
Phenotype
MIM: 607071 MIM: 604038 MIM: 602725 MIM: 607073
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
HYAL1 PubMed Links

Gene Details

Gene
HYAL1
Gene Name
hyaluronoglucosaminidase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033159.3 1771 Silent Mutation GCC,GCG A311A NP_149349.2
NM_153281.1 1771 Silent Mutation GCC,GCG A311A NP_695013.1
NM_153282.2 1771 Intron NP_695014.1
NM_153283.2 1771 Silent Mutation GCC,GCG A129A NP_695015.1
NM_153285.2 1771 Silent Mutation GCC,GCG A52A NP_695017.1
XM_011533667.2 1771 Silent Mutation GCC,GCG A311A XP_011531969.1
XM_011533668.2 1771 Silent Mutation GCC,GCG A311A XP_011531970.1
XM_011533669.2 1771 Silent Mutation GCC,GCG A311A XP_011531971.1
Gene
HYAL3
Gene Name
hyaluronoglucosaminidase 3
There are no transcripts associated with this gene.

Gene
IFRD2
Gene Name
interferon related developmental regulator 2
There are no transcripts associated with this gene.

Gene
NAT6
Gene Name
N-acetyltransferase 6
There are no transcripts associated with this gene.

View Full Product Details